missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Axygen™ PCR Tube Storage Racks

Constructed of polypropylene. Corning™ Axygen™ PCR Tube Storage Racks hold 96 0.2mL PCR tubes. Supplied with lids.

$531.94 - $532.38

Specifications

Product Type Microtube Rack
Includes Clear Polycarbonate Cover
Autoclavable Not Autoclavable
Holds Microtubes (0.2 to 0.5 mL)
Material Polycarbonate
View More Specs

Products 2
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
14-222-397
View Documents
Axygen™
R96PCRFY
Fluorescent Yellow
Case of 25 Each for $532.38
 
14-222-396
View Documents
Axygen™
R96PCRFSP
Assorted (Fluorescent Blue, Green, Red, Violet, Yellow)
Case of 25 Each for $531.94
 
Description

Description

Polypropylene rack for PCR Tubes
  • Holds 96 0.2mL PCR tubes
  • Available in several colors
  • Supplied with lids
  • Packaged five per pack and five packs per case. Spectrum pack includes five racks of each fluorescent color
Specifications

Specifications

Microtube Rack
Not Autoclavable
Polycarbonate
25/Cs.
Clear Polycarbonate Cover
Microtubes (0.2 to 0.5 mL)
96
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'