Learn More
Worthington Biochemical PDS KIT 5/PK DFS Item
Supplier:  Worthington Biochemical LK003178
Papain Dissociation System, Papain Vial, Code: PAP2, Vial reconstituted with 5mL of EBSS or equivalent yields a solution at 20U of papain permL in 1mm L-cysteine with 0.5mm EDTA, Store at 2 deg. to 8 deg.C, 5Vi
Catalog No. NC9212788
Only null left
Explore available promotions