missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cytiva PEEK Tubing

PEEK tubing comes in a variety of internal diameter sizes. Cytiva Lifescience™ PEEK Tubing is an important chromatography accessory.

$162.00 - $162.00

Specifications

Type Tubing
Diameter (Metric) Outer 1.58 mm
Overall Length (Metric) 2 m
Material PEEK
View More Specs

Products 1
Catalog Number Mfr. No. Diameter (Metric) Inner Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Inner Price Quantity  
36-104-1170
View Documents
Cytiva
18111253
0.75 mm
Each for $162.00
 
Description

Description

  • Polyetheretherketone (PEEK)
  • A variety of sizes are available
Specifications

Specifications

Tubing
1.58 mm
2 m
PEEK
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'