missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Pelican™ HeadsUp™ Lites Headlamps
Hands-free light when and where you need it
Supplier: Pelican™ 2610
Includes: 2610 and 2620 Models include three AAA batteries; 2680 Model supplied with four AA batteries.
Description
- Adjustable cloth strap fits comfortably and securely on head
- Rubber strap grips tightly to hardhats and helmets
2610 Model
- 47-hour burn time on low; 19 hours on high
- Dual-mode button activates one or all three LEDs for full brightness
- Length: 1.5 in. (3.8cm)
- Weight (with batteries): 3.2 oz. (90.1g)
- Powered by three “AAA” alkaline cells (included)
- 18-hour burn time on low; 2.25 hours on high
- Hybrid dual lamps; Xenon for intense bright light or three LEDs
- Multi-angle pivoting head allows light to be aimed where needed
- Length: 2.2 in. (5.5cm)
- Weight (with batteries): 4.8 oz. (136kg)
- Powered by three “AAA” alkaline cells (included)
- Outfitted with Recoil LED module
- Uses a 1 watt LED aimed backwards into the reflector
- 100% of the light is captured and aimed forward as a tight collimated beam as bright as an incandescent
- Body is made of tough, corrosion proof, submersible, and virtually indestructible ABS resin
- Powered by four “AA” alkaline cells (included)
- Comes with a rubber strap for hard hat use and an adjustable cloth strap for bare head
- Impact and chemical-resistant ABS'water resistant for all-weather use
- Provides 60 lumens of LED light
- 10 hours of battery burn time
- With rotary bezel switch
- Adjustable cloth strap; heavy-duty rubber strap
- Powered by three “AAA” alkaline cells (included)
Certifications
2610, 2680, and 2690 Models are UL listed for certain Class I, Division 1 hazardous locations; CE marked. 2680 Model is UL listed for certain Class I, Division 2 hazardous locations; CE marked.
Specifications
Headlamp | |
4.5 V | |
0.09 kg | |
Alkaline | |
Black | |
3 AAA alkaline batteries, adjustable cloth strap and rubber strap | |
3.8 cm | |
ABS Resin |
LED | |
3.2 oz. | |
19 hr. | |
UL, Class I, Division 2, CE | |
Hands free light when and where you need it, adjustable cloth strap fits comfortably and securely on head | |
1.49 in. | |
30 lm | |
Lifetime |