missing translation for 'onlineSavingsMsg'
Learn More

Penicillin G sodium salt

Penicillin G sodium salt, CAS # 69-57-8, also called benzylpenicillin, is a β-lactam antibiotic active against gram-positive and gram-negative bacteria. | CAS: 69-57-8 | C16H17N2NaO4S | 356.372 g/mol

$60.38 - $254.21

Chemical Identifiers

CAS 69-57-8
Molecular Formula C16H17N2NaO4S
Molecular Weight (g/mol) 356.372
MDL Number MFCD00069666
InChI Key FCPVYOBCFFNJFS-LQDWTQKMSA-M
Synonym Benzylpenicillin sodium salt
PubChem CID 23668834
ChEBI CHEBI:51765
IUPAC Name sodium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMILES CC1(C(N2C(S1)C(C2=O)NC(=O)CC3=CC=CC=C3)C(=O)[O-])C.[Na+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6303206
View Documents
Thermo Scientific Chemicals
J6303206
5 g N/A
Only null left
 
AAJ6303214
View Documents
Thermo Scientific Chemicals
J6303214
25 g N/A
Only null left
 
AAJ6303222
View Documents
Thermo Scientific Chemicals
J6303222
100 g N/A
Only null left
 
Description

Description

Penicillin G sodium salt is used as a β-lactam antibiotic. Penicillin G can be used as a selective agent in several types of isolation media. It has been used to study the diagnostic and therapeutic implications of gentamicin-resistant Enterococcus faecalis sequence type 6 with reduced penicillin susceptibility and in cell culture both alone and combined with streptomycin and other antibiotics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Penicillin G sodium salt is the sodium salt of benzylpenicillin, a broad-spectrum β-lactam antibiotic with gram-positive and gram-negative bactericidal activity
  • Penicillin G binds and inactivates penicillin-binding-proteins associated with the bacterial cell wall, thus inhibiting cell wall synthesis and resulting in cell lysis

Applications

  • Penicillin G can be used as a selection agent in the isolation of microorganisms or in cell culture applications
  • It can also be used to prevent infection in research animals
Specifications

Chemical Identifiers

69-57-8
356.372
FCPVYOBCFFNJFS-LQDWTQKMSA-M
23668834
sodium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
C16H17N2NaO4S
MFCD00069666
Benzylpenicillin sodium salt
CHEBI:51765
CC1(C(N2C(S1)C(C2=O)NC(=O)CC3=CC=CC=C3)C(=O)[O-])C.[Na+]

Specifications

Penicillin G Sodium Salt
69-57-8
5 g
MFCD00069666
14,7094
Soluble in water (100mg/ml),alcohol,glycerol,DMSO (71mg/ml at 25°C),and ethanol (< 1mg/ml at 25°C).
CC1(C(N2C(S1)C(C2=O)NC(=O)CC3=CC=CC=C3)C(=O)[O-])C.[Na+]
356.372
CHEBI:51765
Penicillin G sodium salt
Powder
White
C16H17N2NaO4S
3834217
Benzylpenicillin sodium salt
FCPVYOBCFFNJFS-LQDWTQKMSA-M
sodium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
23668834
356.37
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c

H317-H334

EINECSNumber : 200-710-2

RTECSNumber : XH9800000

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only