missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentabromobenzyl Alcohol 98.0+%, TCI America™
$65.24 - $180.81
Chemical Identifiers
| CAS | 79415-41-1 |
|---|---|
| Molecular Formula | C7H3Br5O |
| Molecular Weight (g/mol) | 502.62 |
| MDL Number | MFCD00192377 |
| InChI Key | KKWHDMUCBWSKGL-UHFFFAOYSA-N |
| Synonym | 2,3,4,5,6-pentabromobenzyl alcohol, pentabromobenzyl alcohol, 2,3,4,5,6-pentabromophenyl methanol, acmc-1beji, 2,3,4,5,6-pentabromobenzylalcohol, kkwhdmucbwskgl-uhfffaoysa, benzenemethanol,2,3,4,5,6-pentabromo |
| PubChem CID | 2733946 |
| IUPAC Name | (2,3,4,5,6-pentabromophenyl)methanol |
| SMILES | C(C1=C(C(=C(C(=C1Br)Br)Br)Br)Br)O |
Chemical Identifiers
| 79415-41-1 | |
| 502.62 | |
| KKWHDMUCBWSKGL-UHFFFAOYSA-N | |
| 2733946 | |
| C(C1=C(C(=C(C(=C1Br)Br)Br)Br)Br)O |
| C7H3Br5O | |
| MFCD00192377 | |
| 2,3,4,5,6-pentabromobenzyl alcohol, pentabromobenzyl alcohol, 2,3,4,5,6-pentabromophenyl methanol, acmc-1beji, 2,3,4,5,6-pentabromobenzylalcohol, kkwhdmucbwskgl-uhfffaoysa, benzenemethanol,2,3,4,5,6-pentabromo | |
| (2,3,4,5,6-pentabromophenyl)methanol |
Specifications
| 79415-41-1 | |
| C7H3Br5O | |
| 1 g | |
| KKWHDMUCBWSKGL-UHFFFAOYSA-N | |
| (2,3,4,5,6-pentabromophenyl)methanol | |
| 2733946 | |
| ≥98.0% (GC) | |
| Pentabromobenzyl Alcohol |
| Yellow | |
| MFCD00192377 | |
| 2,3,4,5,6-pentabromobenzyl alcohol, pentabromobenzyl alcohol, 2,3,4,5,6-pentabromophenyl methanol, acmc-1beji, 2,3,4,5,6-pentabromobenzylalcohol, kkwhdmucbwskgl-uhfffaoysa, benzenemethanol,2,3,4,5,6-pentabromo | |
| C(C1=C(C(=C(C(=C1Br)Br)Br)Br)Br)O | |
| 502.62 | |
| 502.62 | |
| Crystalline Powder |
Safety and Handling
TSCA : No