missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pentadecanoic acid, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1466418

 View more versions of this product

Catalog No. AAA1466418



Description

Description

Pentadecanoic acid is a lipid used in proteomics research. It is also used as a biomarker for dietary milk intake. It can be labelled as a biomarker for myocardial fatty acid metabolism in ischemia/reperfusion studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentadecanoic acid is a lipid used in proteomics research. It is also used as a biomarker for dietary milk intake. It can be labelled as a biomarker for myocardial fatty acid metabolism in ischemia/reperfusion studies.

Solubility
Soluble in ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from bases, oxidizing agents, reducing agents.
Specifications

Specifications

Pentadecanoic acid
51°C to 54°C
110°C (230°F)
CH3(CH2)13CO2H
50 g
pentadecylic acid, n-pentadecanoic acid, pentadecyclic acid, n-pentadecylic acid, unii-ccw02d961f, pentadecyclate, n-pentadecylate, n-pentadecanoate, n-pentadecanoicacid, pentadecansäure
WQEPLUUGTLDZJY-UHFFFAOYSA-N
pentadecanoic acid
13849
242.4
1002-84-2
257°C (100 mmHg)
C15H30O2
MFCD00002745
1773831
Soluble in ethanol.
CCCCCCCCCCCCCCC(=O)O
242.403
CHEBI:42504
99%

Chemical Identifiers

1002-84-2
242.403
WQEPLUUGTLDZJY-UHFFFAOYSA-N
13849
pentadecanoic acid
C15H30O2
MFCD00002745
pentadecylic acid, n-pentadecanoic acid, pentadecyclic acid, n-pentadecylic acid, unii-ccw02d961f, pentadecyclate, n-pentadecylate, n-pentadecanoate, n-pentadecanoicacid, pentadecansäure
CHEBI:42504
CCCCCCCCCCCCCCC(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 213-693-1

RTECSNumber : RZ1925000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only