Learn More
Pentadecanoic acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1466430
Description
Pentadecanoic acid is a lipid used in proteomics research. It is also used as a biomarker for dietary milk intake. It can be labelled as a biomarker for myocardial fatty acid metabolism in ischemia/reperfusion studies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPentadecanoic acid is a lipid used in proteomics research. It is also used as a biomarker for dietary milk intake. It can be labelled as a biomarker for myocardial fatty acid metabolism in ischemia/reperfusion studies.
Solubility
Soluble in ethanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from bases, oxidizing agents, reducing agents.
Specifications
Pentadecanoic acid | |
51°C to 54°C | |
110°C (230°F) | |
CH3(CH2)13CO2H | |
250 g | |
pentadecylic acid, n-pentadecanoic acid, pentadecyclic acid, n-pentadecylic acid, unii-ccw02d961f, pentadecyclate, n-pentadecylate, n-pentadecanoate, n-pentadecanoicacid, pentadecansäure | |
WQEPLUUGTLDZJY-UHFFFAOYSA-N | |
pentadecanoic acid | |
13849 | |
242.4 |
1002-84-2 | |
257°C (100 mmHg) | |
C15H30O2 | |
MFCD00002745 | |
1773831 | |
Soluble in ethanol. | |
CCCCCCCCCCCCCCC(=O)O | |
242.403 | |
CHEBI:42504 | |
99% |
Chemical Identifiers
1002-84-2 | |
242.403 | |
WQEPLUUGTLDZJY-UHFFFAOYSA-N | |
13849 | |
pentadecanoic acid |
C15H30O2 | |
MFCD00002745 | |
pentadecylic acid, n-pentadecanoic acid, pentadecyclic acid, n-pentadecylic acid, unii-ccw02d961f, pentadecyclate, n-pentadecylate, n-pentadecanoate, n-pentadecanoicacid, pentadecansäure | |
CHEBI:42504 | |
CCCCCCCCCCCCCCC(=O)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 213-693-1
RTECSNumber : RZ1925000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only