missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentadecyl Acetate 95.0+%, TCI America™
$137.94 - $354.17
Chemical Identifiers
| CAS | 629-58-3 |
|---|---|
| Molecular Formula | C17H34O2 |
| Molecular Weight (g/mol) | 270.457 |
| MDL Number | MFCD00056204 |
| InChI Key | AAAIZLQILBFRTJ-UHFFFAOYSA-N |
| Synonym | Acetic Acid Pentadecyl Ester |
| PubChem CID | 522077 |
| IUPAC Name | pentadecyl acetate |
| SMILES | CCCCCCCCCCCCCCCOC(=O)C |
Chemical Identifiers
| 629-58-3 | |
| 270.457 | |
| AAAIZLQILBFRTJ-UHFFFAOYSA-N | |
| 522077 | |
| CCCCCCCCCCCCCCCOC(=O)C |
| C17H34O2 | |
| MFCD00056204 | |
| Acetic Acid Pentadecyl Ester | |
| pentadecyl acetate |
Specifications
| 629-58-3 | |
| Colorless | |
| C17H34O2 | |
| 5 g | |
| AAAIZLQILBFRTJ-UHFFFAOYSA-N | |
| pentadecyl acetate | |
| 522077 | |
| ≥95.0% (GC) | |
| Pentadecyl Acetate |
| 15°C | |
| 230°C | |
| MFCD00056204 | |
| Acetic Acid Pentadecyl Ester | |
| CCCCCCCCCCCCCCCOC(=O)C | |
| 270.457 | |
| 270.46 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0735
TSCA : No