missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Pentafluoro(phenoxy)cyclotriphosphazene 98.0+%, TCI America™
Supplier: TCI America P22861G
Specifications
Pentafluoro(phenoxy)cyclotriphosphazene | |
Colorless | |
C6H5F5N3OP3 | |
1760 | |
XNZZEQCBAGUFMT-UHFFFAOYSA-N | |
2,2,4,4,6-pentafluoro-6-phenoxy-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene | |
23419251 | |
≥98.0% (GC) |
33027-68-8 | |
46°C | |
1 g | |
2,4,4,6,6-Pentafluoro-2-phenoxy-1,3,5,2,4,6-triazatriphosphorine | |
C1=CC=C(C=C1)OP2(=NP(=NP(=N2)(F)F)(F)F)F | |
323.039 | |
323.04 | |
Liquid |
Chemical Identifiers
33027-68-8 | |
323.039 | |
2,4,4,6,6-Pentafluoro-2-phenoxy-1,3,5,2,4,6-triazatriphosphorine | |
2,2,4,4,6-pentafluoro-6-phenoxy-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene |
C6H5F5N3OP3 | |
XNZZEQCBAGUFMT-UHFFFAOYSA-N | |
23419251 | |
C1=CC=C(C=C1)OP2(=NP(=NP(=N2)(F)F)(F)F)F |
Safety and Handling
EINECSNumber : (5)-6917
TSCA : No
Recommended Storage : Refrigerator