missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Pentafluorophenyl)diphenylphosphine 93.0+%, TCI America™
$281.48 - $818.32
Chemical Identifiers
| CAS | 5525-95-1 |
|---|---|
| Molecular Formula | C18H10F5P |
| Molecular Weight (g/mol) | 352.24 |
| MDL Number | MFCD00000290 |
| InChI Key | KUTXTUCJQJPJBH-UHFFFAOYSA-N |
| Synonym | diphenyl pentafluorophenyl phosphine, pentafluorophenyl diphenylphosphine, perfluorophenyl diphenylphosphine, phosphine, pentafluorophenyl diphenyl, pentafluorophenyl diphenylphosphane, pubchem6416, acmc-1aogr, pentafluorodiphenylphosphine, diphenylpentafluorophenyphosphine, pentafluorophenyldiphenylphosphine |
| PubChem CID | 300563 |
| IUPAC Name | (2,3,4,5,6-pentafluorophenyl)diphenylphosphane |
| SMILES | FC1=C(F)C(F)=C(P(C2=CC=CC=C2)C2=CC=CC=C2)C(F)=C1F |
Chemical Identifiers
| 5525-95-1 | |
| 352.24 | |
| KUTXTUCJQJPJBH-UHFFFAOYSA-N | |
| 300563 | |
| FC1=C(F)C(F)=C(P(C2=CC=CC=C2)C2=CC=CC=C2)C(F)=C1F |
| C18H10F5P | |
| MFCD00000290 | |
| diphenyl pentafluorophenyl phosphine, pentafluorophenyl diphenylphosphine, perfluorophenyl diphenylphosphine, phosphine, pentafluorophenyl diphenyl, pentafluorophenyl diphenylphosphane, pubchem6416, acmc-1aogr, pentafluorodiphenylphosphine, diphenylpentafluorophenyphosphine, pentafluorophenyldiphenylphosphine | |
| (2,3,4,5,6-pentafluorophenyl)diphenylphosphane |
Specifications
| 5525-95-1 | |
| White | |
| MFCD00000290 | |
| diphenyl pentafluorophenyl phosphine, pentafluorophenyl diphenylphosphine, perfluorophenyl diphenylphosphine, phosphine, pentafluorophenyl diphenyl, pentafluorophenyl diphenylphosphane, pubchem6416, acmc-1aogr, pentafluorodiphenylphosphine, diphenylpentafluorophenyphosphine, pentafluorophenyldiphenylphosphine | |
| FC1=C(F)C(F)=C(P(C2=CC=CC=C2)C2=CC=CC=C2)C(F)=C1F | |
| 352.24 | |
| 352.24 | |
| Crystalline Powder |
| 70°C | |
| C18H10F5P | |
| 1 g | |
| KUTXTUCJQJPJBH-UHFFFAOYSA-N | |
| (2,3,4,5,6-pentafluorophenyl)diphenylphosphane | |
| 300563 | |
| ≥93.0% (T) | |
| (Pentafluorophenyl)diphenylphosphine |
Safety and Handling
TSCA : No