missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Pentafluorophenyl Trifluoroacetate 95.0+%, TCI America™
Supplier: TCI America P189425G
Specifications
Pentafluorophenyl Trifluoroacetate | |
4°C | |
122°C | |
MFCD00134438 | |
3272 | |
VCQURUZYYSOUHP-UHFFFAOYSA-N | |
2,3,4,5,6-pentafluorophenyl 2,2,2-trifluoroacetate | |
4327891 | |
≥95.0% (GC) |
14533-84-7 | |
Yellow | |
C8F8O2 | |
25 g | |
pentafluorophenyl trifluoroacetate, perfluorophenyl 2,2,2-trifluoroacetate, trifluoroacetic acid pentafluorophenyl ester, pentafluorphenyl trifluoracetate, pentafluorophenyltrifluoroacetate, pentafluorophenyl 2,2,2-trifluoroacetate, acetic acid, trifluoro-, pentafluorophenyl ester, acetic acid,2,2,2-trifluoro-, 2,3,4,5,6-pentafluorophenyl ester, ambotzrl-1046, acmc-1bui8 | |
FC1=C(F)C(F)=C(OC(=O)C(F)(F)F)C(F)=C1F | |
280.07 | |
280.07 | |
Liquid |
Chemical Identifiers
14533-84-7 | |
280.07 | |
VCQURUZYYSOUHP-UHFFFAOYSA-N | |
4327891 | |
FC1=C(F)C(F)=C(OC(=O)C(F)(F)F)C(F)=C1F |
C8F8O2 | |
MFCD00134438 | |
pentafluorophenyl trifluoroacetate, perfluorophenyl 2,2,2-trifluoroacetate, trifluoroacetic acid pentafluorophenyl ester, pentafluorphenyl trifluoracetate, pentafluorophenyltrifluoroacetate, pentafluorophenyl 2,2,2-trifluoroacetate, acetic acid, trifluoro-, pentafluorophenyl ester, acetic acid,2,2,2-trifluoro-, 2,3,4,5,6-pentafluorophenyl ester, ambotzrl-1046, acmc-1bui8 | |
2,3,4,5,6-pentafluorophenyl 2,2,2-trifluoroacetate |
Safety and Handling
TSCA : No