missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pentafluoropropionic anhydride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 416930050

 View more versions of this product

Catalog No. AC416930050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Acylation reagent
Specifications

Specifications

Pentafluoropropionic anhydride
Colorless
69.0°C
97.5% min. (GC)
C6F10O3
(CF3CF2CO)2O
1.57
XETRHNFRKCNWAJ-UHFFFAOYSA-N
2,2,3,3,3-pentafluoropropanoyl 2,2,3,3,3-pentafluoropropanoate
67742
98%
356-42-3
1.5700g/mL
Authentic
Glass bottle
1.3
5 g
pentafluoropropionic anhydride, perfluoropropionic anhydride, pentafluoropropionic acid anhydride, pfpa, perfluoropropionic acid anhydride, propanoic acid, pentafluoro-, anhydride, unii-0lfe7u11o5, pentafluoropropanoic anhydride, decafluoropropionic anhydride, propionic acid, pentafluoro-, anhydride
C(=O)(C(C(F)(F)F)(F)F)OC(=O)C(C(F)(F)F)(F)F
310.04
310.04
Liquid

Chemical Identifiers

356-42-3
310.04
pentafluoropropionic anhydride, perfluoropropionic anhydride, pentafluoropropionic acid anhydride, pfpa, perfluoropropionic acid anhydride, propanoic acid, pentafluoro-, anhydride, unii-0lfe7u11o5, pentafluoropropanoic anhydride, decafluoropropionic anhydride, propionic acid, pentafluoro-, anhydride
2,2,3,3,3-pentafluoropropanoyl 2,2,3,3,3-pentafluoropropanoate
C6F10O3
XETRHNFRKCNWAJ-UHFFFAOYSA-N
67742
C(=O)(C(C(F)(F)F)(F)F)OC(=O)C(C(F)(F)F)(F)F
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 206-604-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only