missing translation for 'onlineSavingsMsg'
Learn More

Pentane (Certified), Fisher Chemical™

$687.33 - $2445.58

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
O4062-4
SDS View Documents
Fisher Chemical
O40624
4 L Glass Bottle
Each for $687.33
Case of 4 Each for $2,749.32
Only null left
 
O4062-20
SDS View Documents
Fisher Chemical
O406220
20 L Steel Pail
Each for $2,445.58
Only null left
 
Description

Description

Specifications

Chemical Identifiers

109-66-0
72.15
OFBQJSOFQDEBGM-UHFFFAOYSA-N
8003
pentane
C5H12
MFCD00009498
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
CHEBI:37830
CCCCC

Specifications

-130°C
0.626g/cm³
4 L
Glass Bottle
≥98 %
CH3(CH2)3CH3
573mbar at 20°C
1265
OFBQJSOFQDEBGM-UHFFFAOYSA-N
pentane
8003
72.15
Certified
109-66-0
35°C
Pass Test
Liquid
C5H12
MFCD00009498
0.25 mPa/s at 20°C
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
CCCCC
72.15
CHEBI:37830
≥98%
Pentane
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Extremely flammable liquid and vapor. Inhalation may cause central nervous system effects. May cause skin, eye, and respiratory tract irritation. Aspiration hazard if swallowed - can enter lungs and cause damage. Repeated exposure may cause skin dryness or cracking. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention.

NFPA

Health:1
Flammability:4
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid

SDS
Documents

Documents