Learn More
Pentanol, mixture of isomers, 99% (sum of isomers), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H33391AP
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Pentanol | |
94624-12-1 | |
128°C to 137°C | |
C5H12O | |
MFCD00081734 | |
UN1987 | |
AMQJEAYHLZJPGS-UHFFFAOYSA-N | |
pentan-1-ol | |
6276 | |
88.15 |
mixture of isomers | |
0.815 | |
45°C (113°F) | |
1.4102 | |
500 mL | |
1-pentanol, amyl alcohol, n-amyl alcohol, n-pentanol, pentanol, pentyl alcohol, butylcarbinol, 1-pentyl alcohol, amylol, n-butylcarbinol | |
CCCCCO | |
88.15 | |
CHEBI:44884 | |
99% |
Chemical Identifiers
94624-12-1 | |
88.15 | |
AMQJEAYHLZJPGS-UHFFFAOYSA-N | |
6276 | |
pentan-1-ol |
C5H12O | |
MFCD00081734 | |
1-pentanol, amyl alcohol, n-amyl alcohol, n-pentanol, pentanol, pentyl alcohol, butylcarbinol, 1-pentyl alcohol, amylol, n-butylcarbinol | |
CHEBI:44884 | |
CCCCCO |
Safety and Handling
GHS H Statement
H226-H332-H315-H335
Flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
H226-H332-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber : 305-536-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only