Learn More
Thermo Scientific Chemicals Pepstatin A, 98%
Protease inhibitor that acts on pepsin, renin and cathepsin D
Supplier: Thermo Scientific Chemicals J60237MB
Description
Pepstatin A, is used as an irreversible inhibitor of HIV-1, aspartic proteases. Inhibits lysosomal proteases and interferes with autolysosomal digestion when used in combination with E 64d. Pepstatin A also efficiently inhibits HIV-1 protease and Fcγ receptor induction by IFN-γ.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Protease inhibitor that acts on pepsin, renin and cathepsin D | |
98% | |
C34H63N5O9 | |
2201362 | |
FAXGPCHRFPCXOO-JHTFMCFMSA-N | |
3-hydroxy-4-[(2S)-2-{3-hydroxy-6-methyl-4-[(2S)-3-methyl-2-[(2S)-3-methyl-2-(3-methylbutanamido)butanamido]butanamido]heptanamido}propanamido]-6-methylheptanoic acid | |
Protease Inhibitor |
685.90g/mol,685.90 | |
26305-03-3 | |
2°C to 8°C | |
Powder | |
CC(C)CC(NC(=O)[C@H](C)NC(=O)CC(O)C(CC(C)C)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CC(C)C)C(C)C)C(C)C)C(O)CC(O)=O | |
25 mg |
Chemical Identifiers
26305-03-3 |
Safety and Handling
EINECSNumber : 247-600-0
Recommended Storage : Keep cold
RUO – Research Use Only