missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing

Pepstatin A, Fisher BioReagents
Supplier: Fisher BioReagents BP2671250
Description
Pepstatin A is an inhibitor for pepsin, renin, cathepsin D, and other acid proteases.Specifications
Pepstatin A | |
235°C | |
230° to 240°C | |
Amber Glass | |
250 mg | |
pepstatin a | |
CC(C)CC(C(CC(=O)O)O)NC(=O)C(C)NC(=O)CC(C(CC(C)C)NC(=O)C(C(C)C)NC(=O)C(C(C)C)NC(=O)CC(C)C)O | |
685.904 | |
685.9 |
26305-03-3 | |
White | |
Conforms to reference | |
C34H63N5O9 | |
15, 7260 | |
FAXGPCHRFPCXOO-WQILDUGRSA-N | |
(3R,4S)-3-hydroxy-4-[[(2R)-2-[[(3R,4S)-3-hydroxy-6-methyl-4-[[(2S)-3-methyl-2-[[(2S)-3-methyl-2-(3-methylbutanoylamino)butanoyl]amino]butanoyl]amino]heptanoyl]amino]propanoyl]amino]-6-methylheptanoic acid | |
131801262 | |
Powder or Solid |
Chemical Identifiers
26305-03-3 | |
685.904 | |
pepstatin a | |
(3R,4S)-3-hydroxy-4-[[(2R)-2-[[(3R,4S)-3-hydroxy-6-methyl-4-[[(2S)-3-methyl-2-[[(2S)-3-methyl-2-(3-methylbutanoylamino)butanoyl]amino]butanoyl]amino]heptanoyl]amino]propanoyl]amino]-6-methylheptanoic acid |
C34H63N5O9 | |
FAXGPCHRFPCXOO-WQILDUGRSA-N | |
131801262 | |
CC(C)CC(C(CC(=O)O)O)NC(=O)C(C)NC(=O)CC(C(CC(C)C)NC(=O)C(C(C)C)NC(=O)C(C(C)C)NC(=O)CC(C)C)O |
Safety and Handling

Recommended Storage : RT