missing translation for 'onlineSavingsMsg'
Learn More

Perchloric acid, 70%, 99.9985% (metals basis)

Perchloric acid: 70% | CAS: 7601-90-3 | ClHO4 | 100.45 g/mol

$97.45 - $487.22

Chemical Identifiers

CAS 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA10983AA
View Documents
Thermo Scientific Chemicals
010983AA
5 mL
Each for $97.45
Only null left
 
AA10983AD
View Documents
Thermo Scientific Chemicals
010983AD
50 mL
Each for $145.81
Only null left
 
AA10983AK
View Documents
Thermo Scientific Chemicals
010983AK
250 mL
Each for $487.22
Only null left
 
Description

Description

Perchloric acid is used as a precursor for ammonium perchlorate, which is used in rocket fuel. It is useful for etching of liquid crystal displays, etching of chrome, ore extraction and in analytical chemistry. It is used as a strong oxidizer and as a solvent for metals and alloys. It serves as a deproteinization reagent in food analysis. As a dehydrating agent, it is used for the determination of silica in iron and steel and in cement and other silicate materials. It is involved in the separation and determination of copper and other metals and alkali metals such as sodium and potassium. It is also used as an adjunct to increase the reduction potential of cerate salts in volumetric analysis. It finds application in the computer industry for etching liquid crystal displays in critical electronic applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7601-90-3
100.45
VLTRZXGMWDSKGL-UHFFFAOYSA-N
24247
perchloric acid
ClHO4
MFCD00011325
unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
CHEBI:29221
O[Cl](=O)(=O)=O

Specifications

Colorless liquid
°C to 18°C
203°C
Pungent
113°C (235°F)
5 mL
Liquid
unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
VLTRZXGMWDSKGL-UHFFFAOYSA-N
perchloric acid
24247
100.46
White
7601-90-3
1.67
9 hPa (7mm Hg)
99.9985% (metals basis)
ClHO4
MFCD00011325
Miscible with water.
O[Cl](=O)(=O)=O
100.45
CHEBI:29221
Perchloric acid 70%
Videos
Safety and Handling

Safety and Handling

P210-P220-P221-P234-P260-P264b-P270-P271-P280-P283-P304+P340-P305+P351+P338-P306+P360-P310-P330-P331-P353-P363-P370+P378q-P371+P380+P375-P390-P501c

H271-H290-H302-H314-H335-H373

DOTInformation : DOT Class: 5.1, Packaging Group: I

EINECSNumber : 231-512-4

RTECSNumber : SC7500000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only