missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Perchloric Acid, 70% (Certified ACS), Fisher Chemical™

Supplier:  Thermo Fisher Scientific FLA229S8LB

 View more versions of this product

Catalog No. A229S-8LB


Explore available promotions

Description

Description

Specifications

Specifications

0.1
Colorless
7601-90-3,7732-18-5
1.660g/cm³
Pass Test
6.8mmHg at 25°C
approx. 11.7
8 lb.
Liquid
1873
VLTRZXGMWDSKGL-UHFFFAOYSA-N
perchloric acid
24247
100.46
Certified ACS
Perchloric Acid, 70%
-18°C
203°C
1ppm max.
Safe-Cote™ Glass
113°C
69.0 to 72.0 w/w %
ClHO4
MFCD00011325
unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
O[Cl](=O)(=O)=O
100.45
CHEBI:29221
69.0 to 72.0% (w/w)

Chemical Identifiers

7601-90-3
100.45
VLTRZXGMWDSKGL-UHFFFAOYSA-N
24247
perchloric acid
ClHO4
MFCD00011325
unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
CHEBI:29221
O[Cl](=O)(=O)=O
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Use personal protective equipment. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:3
Flammability:0
Instability:3

DOTInformation : DOT Class 5.1, : Oxidizer

SDS Documents

Documents

Promotions

Promotions