missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Perchloric Acid 70% AR (ACS), Macron Fine Chemicals™

macron

Supplier:  Avantor 399946

Catalog No. MK3999212

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Perchloric Acid
SAFEMOR
2.5 L
Liquid
unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
O[Cl](=O)(=O)=O
100.45
CHEBI:29221
ACS
7601-90-3
60%
ClHO4
MFCD00011325
VLTRZXGMWDSKGL-UHFFFAOYSA-N
perchloric acid
24247
100.46
Videos
SDS
Documents

Documents

Promotions

Promotions