missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Perchloric Acid Solution, N/10 (0.1N) (Certified), Fisher Chemical™

Supplier:  Thermo Fisher Scientific SP339500

 View more versions of this product

Catalog No. SP339-500



Explore available promotions

Description

Description

Specifications

Specifications

0.1
Colorless
7601-90-3,64-19-7
1.060g/cm³
Lot Number Used (KHP Standard)
ClHO4
MFCD00011325
VLTRZXGMWDSKGL-UHFFFAOYSA-N
perchloric acid
24247
Certified
Perchloric Acid Solution
N/10 (0.1N)
0.0995 to 0.1005N
Glass Bottle
500 mL
Liquid
unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
O[Cl](=O)(=O)=O
100.45
CHEBI:29221
0.1N (N/10)

Chemical Identifiers

7601-90-3
100.45
VLTRZXGMWDSKGL-UHFFFAOYSA-N
24247
perchloric acid
ClHO4
MFCD00011325
unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
CHEBI:29221
O[Cl](=O)(=O)=O
Videos
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 8, : Corrosive

SDS Documents

Documents

Promotions

Promotions