missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Perchloric acid, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 452850010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Perchloric Acid | |
7601-90-3,7732-18-5 | |
1.6600g/mL | |
113°C | |
3.5 mPa.s (20°C) | |
HClO4 | |
unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4 | |
VLTRZXGMWDSKGL-UHFFFAOYSA-N | |
perchloric acid | |
24247 | |
100.46 |
-18.0°C | |
203.0°C | |
1.66 | |
1 L | |
ClHO4 | |
MFCD00011325 | |
Solubility in water: soluble. | |
O[Cl](=O)(=O)=O | |
100.45 | |
CHEBI:29221 | |
Analytical |
Chemical Identifiers
7601-90-3 | |
100.45 | |
VLTRZXGMWDSKGL-UHFFFAOYSA-N | |
24247 | |
perchloric acid |
ClHO4 | |
MFCD00011325 | |
unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4 | |
CHEBI:29221 | |
O[Cl](=O)(=O)=O |
RUO – Research Use Only