missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Percoll™, MP Biomedicals

$114.85 - $1367.29

Chemical Identifiers

CAS 65455-52-9
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19536925
View Documents
MP Biomedicals Inc
0219536925
25 mL
Each for $114.85
 
ICN19536990
View Documents
MP Biomedicals Inc
0219536990
500 mL
Each for $1,367.29
 
Description

Description

Specifications

Chemical Identifiers

65455-52-9

Specifications

65455-52-9
25 mL
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'