missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Perfex™ Heavy Duty Floor Squeegees

Supplier:  Perfex™ 9518

 View more versions of this product

Catalog No. 19-042-043



Explore available promotions

Requires : Use with 19-042-040 adjustable polymer handle (sold separately).

Description

Description

With two working edges, this PVC squeegee blade is nonconductive, nonmarring and tear resistant. Mounted on a patented self-locking connector for easy handle attachment, the nonsparking blade performs beautifully on abrasive or rough surfaces. The blade and plastic handle are impervious to water, grease, petroleum products, and solvents, and will not absorb bacteria, liquids, or odors.

Order Info

Comes in three convenient widths: 18, 24, and 30 in. (46, 61, and 76cm).

Specifications

Specifications

Floor Squeegee
PVC blade, Polymer handle
46 cm
Abrasive or rough surfaces
18 in.
Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions