missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Perfluoro(2-methyl-2-pentene), 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1839414

 View more versions of this product

Catalog No. AAA1839414


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Perfluoro(2-methyl-2-pentene)
1.62
−12°C (10°F)
ca. 1.3
25 g
perfluoro-2-methyl-2-pentene, perfluoro 2-methylpent-2-ene, hexafluoropropene dimer, perfluoro-2-methylpent-2-ene, 1,1,1,3,4,4,5,5,5-nonafluoro-2-trifluoromethyl pent-2-ene, perfluoro 2-methyl-2-pentene, nonafluoro-2-trifluoromethyl pent-2-ene, 1,1,1,3,4,4,5,5,5-nonafluoro-2-trifluoromethyl pent-4-ene, 2-pentene, 1,1,1,3,4,4,5,5,5-nonafluoro-2-trifluoromethyl, 1,1,1,3,4,4,5,5,5-nonafluoro-2-trifluoromethyl-2-pentene
C(=C(C(C(F)(F)F)(F)F)F)(C(F)(F)F)C(F)(F)F
300.047
300.05
1584-03-8
48°C to 54°C
C6F12
MFCD00015724
UN1993
FAEGGADNHFKDQX-UHFFFAOYSA-N
1,1,1,3,4,4,5,5,5-nonafluoro-2-(trifluoromethyl)pent-2-ene
74105
97%

Chemical Identifiers

1584-03-8
300.047
FAEGGADNHFKDQX-UHFFFAOYSA-N
74105
C(=C(C(C(F)(F)F)(F)F)F)(C(F)(F)F)C(F)(F)F
C6F12
MFCD00015724
perfluoro-2-methyl-2-pentene, perfluoro 2-methylpent-2-ene, hexafluoropropene dimer, perfluoro-2-methylpent-2-ene, 1,1,1,3,4,4,5,5,5-nonafluoro-2-trifluoromethyl pent-2-ene, perfluoro 2-methyl-2-pentene, nonafluoro-2-trifluoromethyl pent-2-ene, 1,1,1,3,4,4,5,5,5-nonafluoro-2-trifluoromethyl pent-4-ene, 2-pentene, 1,1,1,3,4,4,5,5,5-nonafluoro-2-trifluoromethyl, 1,1,1,3,4,4,5,5,5-nonafluoro-2-trifluoromethyl-2-pentene
1,1,1,3,4,4,5,5,5-nonafluoro-2-(trifluoromethyl)pent-2-ene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapour.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 216-436-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only