Learn More
Perfluoro(2-methyl-3-oxahexanoyl) fluoride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1648314
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Perfluoro(2-methyl-3-oxahexanoyl) fluoride | |
54°C to 56°C | |
C6F12O2 | |
MFCD00054657 | |
UN3265 | |
2,3,3,3-tetrafluoro-2-perfluoropropoxy propanoyl fluoride, undecafluoro-2-methyl-3-oxahexanoyl fluoride, perfluoro 2-methyl-3-oxahexanoyl fluoride, 2,3,3,3-tetrafluoro-2-heptafluoropropoxy propanoyl fluoride, perfluoro-2-methyl-3-oxahexanoyl fluoride, propanoyl fluoride, 2,3,3,3-tetrafluoro-2-heptafluoropropoxy, 2,3,3,3-tetrafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propanoyl fluoride, propanoyl fluoride, 2,3,3,3-tetrafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy, hfpo dimer, acmc-1cljd | |
C(=O)(C(C(F)(F)F)(OC(C(C(F)(F)F)(F)F)(F)F)F)F | |
332.045 | |
332.06 |
2062-98-8 | |
None | |
1.3 | |
25 g | |
Moisture sensitive | |
BCLQALQSEBVVAD-UHFFFAOYSA-N | |
2,3,3,3-tetrafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propanoyl fluoride | |
102740 | |
97% |
Chemical Identifiers
2062-98-8 | |
332.045 | |
BCLQALQSEBVVAD-UHFFFAOYSA-N | |
102740 | |
C(=O)(C(C(F)(F)F)(OC(C(C(F)(F)F)(F)F)(F)F)F)F |
C6F12O2 | |
MFCD00054657 | |
2,3,3,3-tetrafluoro-2-perfluoropropoxy propanoyl fluoride, undecafluoro-2-methyl-3-oxahexanoyl fluoride, perfluoro 2-methyl-3-oxahexanoyl fluoride, 2,3,3,3-tetrafluoro-2-heptafluoropropoxy propanoyl fluoride, perfluoro-2-methyl-3-oxahexanoyl fluoride, propanoyl fluoride, 2,3,3,3-tetrafluoro-2-heptafluoropropoxy, 2,3,3,3-tetrafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propanoyl fluoride, propanoyl fluoride, 2,3,3,3-tetrafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy, hfpo dimer, acmc-1cljd | |
2,3,3,3-tetrafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propanoyl fluoride |
Safety and Handling
GHS H Statement
H314-H318-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 218-173-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only