missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA) Acylation Reagents
Prepare volatile and stable fluoracyl derivatives for GC/MS with the Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA).
$703.92 - $2095.26
Specifications
Storage | Room Temperature |
---|---|
For Use With | GC/MS |
Standard Concentration | 98 to 100% |
Catalog Number | Mfr. No. | Standard Components | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Standard Components | Quantity | Price | Quantity | ||||
PI63163
|
Thermo Scientific™
TS63163 |
Heptafluoro-butanoic acid anhydride | 25 g |
Pack for $719.21
|
|
||||
PI63164
|
Thermo Scientific™
TS63164 |
Heptafluoro-butanoic acid anhydride | 10 x 1 mL |
Pack of 10 Each for $703.92
|
|
||||
PI65191
|
Thermo Scientific™
TS65191 |
Pentafluoro-propanoic acid anhydride | 100 g |
Each for $1,570.02
|
|
||||
PI67363
|
Thermo Scientific™
TS67363 |
Trifluoroacetic acid anhydride | 100 g |
Pack for $2,095.26
|
|
||||
Description
Prepare fluoracyl derivatives for GC/MS and for flame ionization and electron-capture detection techniques with the Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA). These reagents react readily with alcohols, phenols, and amines.
- Used to prepare fluoracyl derivatives for GC/MS
- Produce stable volatile derivatives for FID and ECD techniques
Specifications
Room Temperature | |
GC/MS | |
98 to 100% |
For Research Use Only. Not for use in diagnostic procedures.