Learn More
Perfluoro(decahydronaphthalene), cis + trans, 95%, Thermo Scientific Chemicals
$89.93 - $1,311.58
Chemical Identifiers
CAS | 306-94-5 |
---|---|
Molecular Formula | C10F18 |
Molecular Weight (g/mol) | 462.081 |
MDL Number | MFCD00010626 |
InChI Key | UWEYRJFJVCLAGH-UHFFFAOYSA-N |
Synonym | perfluorodecalin, perflunafene, octadecafluorodecahydronaphthalene, cis-perfluorodecalin, trans-perfluorodecalin, perfluorodecahydronaphthalene, naphthalene, octadecafluorodecahydro, f-dc, pp 5, perflunafenum latin |
PubChem CID | 9386 |
ChEBI | CHEBI:38848 |
IUPAC Name | 1,1,2,2,3,3,4,4,4a,5,5,6,6,7,7,8,8,8a-octadecafluoronaphthalene |
SMILES | C12(C(C(C(C(C1(F)F)(F)F)(F)F)(F)F)(C(C(C(C2(F)F)(F)F)(F)F)(F)F)F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1828814
|
Thermo Scientific Chemicals
A1828814 |
25 g |
N/A
|
|
|||||
AAA1828822
|
Thermo Scientific Chemicals
A1828822 |
100 g |
N/A
|
|
|||||
AAA1828836
|
Thermo Scientific Chemicals
A1828836 |
500 g |
N/A
|
|
|||||
Description
Perfluoro(decahydronaphthalene) is used to dissolve gases. Due to its gas carrying capacity, it is utilized to increase oxygen delivery during cell culture. It acts as an ingredient in fluosol which is an artificial blood product. It is also used to dissolve Teflon.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPerfluoro(decahydronaphthalene) is used to dissolve gases. Due to its gas carrying capacity, it is utilized to increase oxygen delivery during cell culture. It acts as an ingredient in fluosol which is an artificial blood product. It is also used to dissolve Teflon.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
306-94-5 | |
462.081 | |
UWEYRJFJVCLAGH-UHFFFAOYSA-N | |
9386 | |
1,1,2,2,3,3,4,4,4a,5,5,6,6,7,7,8,8,8a-octadecafluoronaphthalene |
C10F18 | |
MFCD00010626 | |
perfluorodecalin, perflunafene, octadecafluorodecahydronaphthalene, cis-perfluorodecalin, trans-perfluorodecalin, perfluorodecahydronaphthalene, naphthalene, octadecafluorodecahydro, f-dc, pp 5, perflunafenum latin | |
CHEBI:38848 | |
C12(C(C(C(C(C1(F)F)(F)F)(F)F)(F)F)(C(C(C(C2(F)F)(F)F)(F)F)(F)F)F)F |
Specifications
306-94-5 | |
1.94 | |
C10F18 | |
MFCD00010626 | |
2067113 | |
perfluorodecalin, perflunafene, octadecafluorodecahydronaphthalene, cis-perfluorodecalin, trans-perfluorodecalin, perfluorodecahydronaphthalene, naphthalene, octadecafluorodecahydro, f-dc, pp 5, perflunafenum latin | |
UWEYRJFJVCLAGH-UHFFFAOYSA-N | |
1,1,2,2,3,3,4,4,4a,5,5,6,6,7,7,8,8,8a-octadecafluoronaphthalene | |
9386 | |
462.1 | |
Perfluoro(decahydronaphthalene), cis + trans |
-10°C to -7°C | |
141°C to 142°C | |
1.3145 | |
25 g | |
14,4183 | |
Insoluble in water. | |
C12(C(C(C(C(C1(F)F)(F)F)(F)F)(F)F)(C(C(C(C2(F)F)(F)F)(F)F)(F)F)F)F | |
462.081 | |
CHEBI:38848 | |
95% |
Safety and Handling
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Hazard Class: 3; Packaging Group: III
EINECSNumber : 206-192-4
RTECSNumber : QJ3175000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only