missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Perfluorobutanoic Acid Analytical Standard, MilliporeSigma™ Supelco™
Perfluorobutanoic Acid is an environmental pollutant, belonging to the perfluorochemical (PFCs) class of compounds. These compounds are widely used in a variety of industrial and commercial applications such as fire-fighting foams, paper, leather and textile coatings, polymer industries, etc.
Supplier: Merck Emd Millipore 6880825MG
Specifications
1.645 g/mL (at 25°C (literature)) | |
C4HF7O2 | |
n20/D 1.3 (literature) | |
25 mg | |
7 (vs air) | |
∼10 mmHg (25°C) | |
Analytical Standard |
120°C (755 mmHg) (literature) | |
CF3CF2CF2COOH | |
MFCD00004171 | |
Heptafluorobutyric acid; Edman Reagent No. 3; HFBA; Perfluorobutyric acid | |
214.04 | |
≥98% (GC) |