Contec™ Peridox™ Concentrate Disinfectant
Peridox™ Concentrate is an EPA-registered disinfectant and cleaner that was designed to eliminate and prevent the spread of spores, as well as more than 40 other pathogens.
$94.57 - $94.57
Specifications
Sterility | Non-sterile |
---|---|
For Use With (Application) | Designed to eliminate and prevent the spread of spores, as well as more than 40 other pathogens |
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
19-165-903
|
Contec™
SPTS100PAA |
Test Strips |
Pack of 100 Each for $94.57
|
|
|||||
Description
- Broad spectrum disinfectant is fast acting and is effective against bacterial spores in 3 minutes, bacteria and viruses in 2 minutes, fungus in 1 minute and TB in 5 minutes.
- Has more than 40 EPA registered claims
- Does not contain bleach, alcohol, phenols or quaternary ammonium compounds.
- Fast acting and will not damage stainless steel equipment
- Compatible with all Contec mopping systems and hardware
- Requires no rinsing and leaves no film on surfaces making it ideal for use on critical surfaces
- With its shorter contact times and powerful wetting agents, Peridox Concentrate stays wet on the surface longer and works faster with shorter contact times and powerful wetting agents
- Will not damage clothing, carpet, upholstery, LCD screens, and other surfaces when used as directed.
- Patented, hyperactive chemistry provides fast results, 3-minute sporicide claim at 99.9999% efficacy
- EPA-registered disinfectant and cleaner is ideal for use in life science cleanrooms
- Concentrated formula provides cleanroom facilities with a cost-effective approach to use a high-level sporicidal disinfectant facility-wide
- Compatible with most cleanroom materials of construction, and can be used on workstations, stainless steel equipment, floors, walls, and ceilings
Specifications
Non-sterile | |
Designed to eliminate and prevent the spread of spores, as well as more than 40 other pathogens |