missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Contec™ PeridoxRTU™ Sporicidal Disinfectant
PeridoxRTU ™ is a simple one-step cleaner, disinfectant and sporicide.
Supplier: Contec™ CR85366IR
Description
- Broad spectrum disinfectant effective against bacterial spores, bacteria, viruses, TB and fungi (mold).
- Excellent material compatibility that will not damage most surfaces in cleanroom environments, including stainless steel, acrylic, glass, aluminum and chrome.
- Does not contain alcohol, bleach, quat or silver.
- Daily cleaning and disinfecting tasks, such as wiping surfaces and mopping floors.
- Effective component to monthly cleaning protocols.
- PeridoxRTU is ideal for use in pharmaceutical, biopharmaceutical and medical device facilities.
- Sterile PeridoxRTU is ideal for pharmaceutical and life science cleanroom environments, including sterile core areas.
- Wear appropriate gloves and eye protection. Additional PPE may be required based on engineering controls/environment, method of application, and use protocols.
Specifications
1.9 to 2.2 (at 20°C) | |
Store in a cool, dry, well ventilated area away from sources of heat, combustible materials, direct sunlight, and incompatible substances. Keep container tightly closed when not in use. Maintain temperature between 0°C and 40°C. Do not allow product to freeze or become overheated. This may cause increased degradation of the product, which will decrease product effectiveness. | |
Ready to use. Do not dilute. | |
1 gal. |
Sterile | |
Double bagged | |
Consult with the US EPA Guidelines listed in 40 CFR Part 261.3 or the EU Directive 2008/98/EC on waste for the classifications of hazardous waste prior to disposal. Furthermore, consult with your state, local, or provincial waste requirements or guidelines, if applicable, to ensure compliance. Arrange disposal in accordance to the EPA and/or state and local guidelines. |