Learn More


Periodic Acid (Certified) ACS, Fisher Chemical™
Supplier: Fisher Chemical FLA223100
Specifications
2 | |
White-Yellow | |
1.4 g/cm3 | |
Glass Bottle | |
≥99% | |
H5IO6 | |
MFCD00011344 | |
orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst | |
O[I](O)(O)(O)(O)=O | |
227.94 | |
CHEBI:29150 | |
≥99% |
Periodic Acid | |
10450-60-9 | |
Pass Test | |
122°C | |
100 g | |
Solid | |
3085 | |
TWLXDPFBEPBAQB-UHFFFAOYSA-N | |
tetrahydroxyiodous acid | |
25289 | |
227.96 | |
Certified |
Chemical Identifiers
10450-60-9 | |
227.94 | |
TWLXDPFBEPBAQB-UHFFFAOYSA-N | |
25289 | |
tetrahydroxyiodous acid |
H5IO6 | |
MFCD00011344 | |
orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst | |
CHEBI:29150 | |
O[I](O)(O)(O)(O)=O |
Safety and Handling


DANGER!
Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Use personal protective equipment. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.
NFPA
Health:3
Flammability:0
Instability:2
DOTInformation : DOT Class 5.1, : Oxidizer