missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Periodic Acid (Certified) ACS, Fisher Chemical™

Supplier:  Fisher Chemical FLA22325

 View more versions of this product

Catalog No. A223-25


Explore available promotions

Description

Description

Specifications

Specifications

2
White-Yellow
1.4 g/cm3
Glass Bottle
≥99%
H5IO6
MFCD00011344
orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst
O[I](O)(O)(O)(O)=O
227.94
CHEBI:29150
≥99%
Periodic Acid
10450-60-9
Pass Test
122°C
25 g
Solid
3085
TWLXDPFBEPBAQB-UHFFFAOYSA-N
tetrahydroxyiodous acid
25289
227.96
Certified

Chemical Identifiers

10450-60-9
227.94
TWLXDPFBEPBAQB-UHFFFAOYSA-N
25289
tetrahydroxyiodous acid
H5IO6
MFCD00011344
orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst
CHEBI:29150
O[I](O)(O)(O)(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Use personal protective equipment. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:3
Flammability:0
Instability:2

DOTInformation : DOT Class 5.1, : Oxidizer

SDS Documents

Documents

Promotions

Promotions