Poltex 6 Pocket PETG Serological Pipette Holder DFS Item
The PETG Serological Pipette Holder is available in 3 pocket capacities, with 3 mounting options, and the option to add a pipette filler bracket to any version.
Supplier:  Poltex PGSERPIP6M
 View more versions of this product
Catalog No. 22-000-620
Explore available promotions
Description
Description
  • Serological Pipette (pipet) Holder.
  • Choose from 4, 5, or 6 pocket capacity.
  • Choose from powerful, interchangeable magnet mount, 3M VHB (very high bond) tape mount, or countertop placement.
  • Add a pipette filler bracket to any version (bracket mounts to either side).
  • Magnets and tape mount to either side.
  • Break-resistant PETG construction.
  • PETG has good alcohol/chemical resistance.
  • Shelves are removable for easy cleaning.
Specifications
Specifications
Holder Only
PETG
Magnet Mount
Serological Pipet Holder
6 Pockets