Learn More
Petrolatum, white, pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 417090100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Petrolatum, white | |
50.0°C to 56.0°C | |
0.8400g/mL | |
>150°C | |
Plastic drum | |
MFCD00147839 | |
0.84 | |
1,1,2-trimethyl-1h-benzo e indole, 1,1,2-trimethyl-1h-benz e indole, 1,1,2-trimethylbenz e indole, 2,3,3-trimethyl-4,5-benzo-3h-indole, 2,3,3-trimethylbenzoindolenine, 1,1,2-trimethylbenzo e indole, 1h-benz e indole, 1,1,2-trimethyl, 2,3,3-trimethyl-1h-benz e indolenine, 1,1,2-trimethylhydrobenzo e indole | |
WJZSZXCWMATYFX-UHFFFAOYSA-N | |
1,1,2-trimethylbenzo[e]indole | |
170530 | |
Pure |
8009-03-8 | |
White | |
>300.0°C | |
Authentic | |
C15H15N | |
10 kg | |
14, 7185 | |
Solubility in water: insoluble. Other solubilities: practically insoluble in glycerol, alcohol, soluble in benzene, chloroform, ether, petroleum, ether, carbon disulfide, oils | |
CC1=NC2=C(C1(C)C)C3=CC=CC=C3C=C2 | |
209.29 | |
5-8 mm2/s (100°C) | |
Paste |
Chemical Identifiers
8009-03-8 | |
209.29 | |
WJZSZXCWMATYFX-UHFFFAOYSA-N | |
170530 | |
CC1=NC2=C(C1(C)C)C3=CC=CC=C3C=C2 |
C15H15N | |
MFCD00147839 | |
1,1,2-trimethyl-1h-benzo e indole, 1,1,2-trimethyl-1h-benz e indole, 1,1,2-trimethylbenz e indole, 2,3,3-trimethyl-4,5-benzo-3h-indole, 2,3,3-trimethylbenzoindolenine, 1,1,2-trimethylbenzo e indole, 1h-benz e indole, 1,1,2-trimethyl, 2,3,3-trimethyl-1h-benz e indolenine, 1,1,2-trimethylhydrobenzo e indole | |
1,1,2-trimethylbenzo[e]indole |
Safety and Handling
GHS H Statement:
GHS P Statement:
Wear protective gloves/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 232-373-2
RUO – Research Use Only