missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Petroleum ether, ACS Reagent, Honeywell Riedel-de Haën™

ACS Reagent

Supplier:  Honeywell-Riedel-de Haen 6738384X4L

 View more versions of this product

Catalog No. 6001168


Explore available promotions

Description

Description

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
Specifications

Specifications

Petroleum ether
8032-32-4
0.64 g/mL at 25 °C(lit.)
4 x 4 L
Aluminum Bottle
UN1268
ACS Reagent
ACS Reagent
100.0000%
35°C to 60°C
−18°C
MFCD00081849
p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide

Chemical Identifiers

8032-32-4
MFCD00081849
p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide
Videos
Safety and Handling

Safety and Handling

ShelfLife : 547 days from date of manufacture

DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Petroleum distillates, n.o.s.

EINECSNumber : 232-453-7

SDS
Documents

Documents

Promotions

Promotions