missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Petroleum Ether, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.0% (C5 & C6 Isomers), Honeywell
Supplier: Honeywell Chemicals LP3174
Specifications
Petroleum Ether | |
8032-32-4 | |
0.64 g/cm3 | |
4 x 4 L | |
Glass Bottle | |
0.02% max. | |
>99% |
ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.0% (C5 & C6 Isomers) | |
100.0000% | |
35°C to 60°C | |
−18°C | |
UN1268 | |
p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide | |
ACS Reagent |
Chemical Identifiers
8032-32-4 | |
p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Petroleum distillates, n.o.s.