missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Petroleum Ether, B.R. 60 DEG -90°C, Spectrum™ Chemical
$1,220.71 - $1,220.71
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18-612-205
|
Spectrum Chemical Mfg Cor
P10504LTGL |
4 L | Amber Glass Bottle |
N/A
|
|
||||
18-612-207
|
Spectrum Chemical Mfg Cor
P1050200LTBL |
200 L | Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
8032-32-4 |
Specifications
8032-32-4 | |
To Pass Test | |
To Pass Test | |
1e-05 |
1 | |
4 L | |
Amber Glass Bottle | |
Reagent |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.