missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Petroleum ether, extra pure, boiling range 60-95°C, Thermo Scientific Chemicals

$122.56 - $815.58

Chemical Identifiers

CAS 64742-49-0
Molecular Formula C6H14
Molecular Weight (g/mol) 86.178
MDL Number MFCD00081849
InChI Key AFABGHUZZDYHJO-UHFFFAOYSA-N
Synonym isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
PubChem CID 7892
SMILES CCCC(C)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC292510010
SDS View Documents
Thermo Scientific Chemicals
292510010
1 L Glass Bottle
Each for $122.56
 
AC292510025
SDS View Documents
Thermo Scientific Chemicals
292510025
2.5 L Glass Bottle
Each for $260.75
 
AC292510100
SDS View Documents
Thermo Scientific Chemicals
292510100
10 L Plastic Drum
Each for $815.58
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

64742-49-0
86.178
AFABGHUZZDYHJO-UHFFFAOYSA-N
7892
C6H14
MFCD00081849
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
CCCC(C)C

Specifications

-40.0°C
0.7000g/mL
1 L
Authentic
Liquid
0.005% max.
C6H14
0.57 mm2/s (25°C)
Solubility in water: insoluble. Other solubilities: miscible with absolute alcohol, benzene,, chloroform, ether, carbon disulfide, carbon, tetrachloride and oils except castor oil.
CCCC(C)C
86.178
Extra Pure
64742-49-0
60.0°C to 95.0°C
−26°C
Glass Bottle
1.3880 to 1.3920 (20°C, 589nm)
0.7
MFCD00081849
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
AFABGHUZZDYHJO-UHFFFAOYSA-N
2-methylpentane
7892
Petroleum ether
Videos
Safety and Handling

Safety and Handling

GHS P Statement
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.

GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only