Learn More
![](http://beta-static.fishersci.ca/images/Solvent.gif)
![](http://beta-static.fishersci.ca/images/Red.gif)
Petroleum Ether, Optima™ for HPLC and GC, Fisher Chemical™
High purity solvent for GC and pesticide residue analysis
Supplier: Thermo Fisher Scientific FLE1204
Description
- Tested by GC-ECD to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
- Also meets ACS specifications
- Intended for research use only
Specifications
Petroleum Ether | |
Colorless | |
38°C | |
Pass Test | |
Liquid | |
1.3560 to 1.3660 (at 25°C) | |
MFCD00081849 | |
0.01% max. | |
OPTIMA™ |
8032-32-4 | |
Pass Test | |
4 L | |
Amber Soda Lime Glass | |
High purity extraction solvent for GC applications | |
1ppm max. | |
1268 | |
p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide |
Chemical Identifiers
MFCD00081849 |
Safety and Handling
![](http://beta-static.fishersci.ca/images/Solvent.gif)
![](http://beta-static.fishersci.ca/images/Red.gif)
DANGER!
Emergency Overview
Flammable liquid and vapor. May cause heritable genetic damage. May cause eye, skin, and respiratory tract irritation. Inhalation may cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.
NFPA
Health:1
Flammability:3
Instability:0
DOTInformation : DOT Class 3, : Flammable Liquid