Promega pGEM™ Vectors DFS Item
Enable blue/white screening and contain both the SP6 and T7 RNA polymerase promoters
Supplier:  Promega P2151
 View more versions of this product
Catalog No. PR-P2151
Explore available promotions
Includes: Vector, 20μg; Bacterial Strain JM109, 500μL
Description
Description
  • Standard Cloning Vector that can Efficiently Synthesize RNA In vitro
  • Blue/white screening to easily identify recombinant clones
  • Versatile vector used for standard cloning and in vitro transcription from SP6 and T7 RNA polymerase promoters flanking the multiple cloning region
  • Multiple cloning site provides a convenient selection of restriction sites for cloning
Specifications
Specifications
SP6, T7
-30°C to -10°C
pGEM-3Z vector
Acc I, Ava I, BamH I, EcoRI, Hinc II, Hind III, Kpn I, Pst I, Sac I, Sal I, Sma I, Sph I, Xba I, Xma I
20 μg
Synthesis of RNA in-vitro and allows selection of recombinants by blue/white screening
Product Suggestions
Product Suggestions