Promega pGEM™ Vectors DFS Item
Enable blue/white screening and contain both the SP6 and T7 RNA polymerase promoters
Supplier:  Promega P2251
 View more versions of this product
Catalog No. PR-P2251
Explore available promotions
Includes: Vector, 20μg; Bacterial Strain JM109, 500μL
Description
Description
  • Standard Cloning Vectors and Templates for In vitro Transcription and Circular ssDNA Production
  • Blue/white screening for easily identifying recombinant clones
  • In vitro transcription from SP6 and T7 RNA polymerase promoters flanking the multiple cloning region
  • Orientation of the f1 origin differentiates vectors (+ and -)
Specifications
Specifications
SP6, T7
-30°C to -10°C
pGEM-7Zf(+) vector
ApaI, AatII, SphI, XbaI, XhoI, EcoRI, KpnI, SmaI, ClaI, HindIII, BamHI, SacI, BstXI and NsiI
20 μg
Synthesis of RNA in-vitro and allows selection of recombinants by blue/white screening