missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
VeriSpec™ pH Buffer, Certified Reference Material
ISO 17025/17034 pH Buffer Certified Reference Materials provide the highest level of quality assurance. A certificate of analysis providing the specified property value, uncertainty, and traceability is available with every bottle.
Supplier: VeriSpec™ RV160100500A
Description
- No formaldehyde or mercury compounds as preservatives
- No chromium compounds as colorants
- Manufactured and tested in ISO 17025/17034 accredited facilities
- pH 4, 7, and 10 are color coded for easy identification
- Certified traceable to NIST
Specifications
VeriSpec™ Certified Reference Material, Buffer pH 10.00 ±0.01 (at 25°C) | |
Blue | |
Yes | |
Calibration | |
10.00 ±0.01 | |
VeriSpec | |
pH Buffer | |
Natural Poly | |
Certified |
Proprietary,7732-18-5,1310-73-2,497-19-8,144-55-8,1310-73-2 | |
Proprietary | |
1X | |
pH Meter and Electrode | |
Liquid | |
500 mL | |
Aqueous Solution | |
NIST Traceable |
ISO 17025/ISO 17034