Ricca VeriSpec™ pH Buffer, Certified Reference Material
Ricca's VeriSpec™ ISO 17025/17034 pH Buffer Certified Reference Materials provide the highest level of quality assurance. A certificate of analysis providing the specified property value, uncertainty, and traceability is available with every bottle.
Supplier:  Ricca VeriSpec™ RV161500500G
 View more versions of this product
Catalog No. 01-185-016
Explore available promotions
Description
Description
  • No formaldehyde or mercury compounds as preservatives
  • No chromium compounds as colorants
  • Manufactured and tested in ISO 17025/17034 accredited facilities
  • pH 4, 7, and 10 are color coded for easy identification
  • Certified traceable to NIST
Specifications
Specifications
7732-18-5,7447-40-7,1310-73-2
Yes
pH Meter and Electrode
Liquid
500 mL
500 mL PETG Clear
Certified
Colorless
Calibration
12.00 ±0.01
VeriSpec
pH Buffer
ISO 17025/17034
Videos
SDS
Documents
Documents
Promotions
Promotions
ISO 17025/ISO 17034