Ricca VeriSpec™ pH Buffer, Certified Reference Material
Ricca's VeriSpec™ ISO 17025/17034 pH Buffer Certified Reference Materials provide the highest level of quality assurance. A certificate of analysis providing the specified property value, uncertainty, and traceability is available with every bottle.
Supplier: Ricca VeriSpec™ RV1500001A
Description
- No formaldehyde or mercury compounds as preservatives
- No chromium compounds as colorants
- Manufactured and tested in ISO 17025/17034 accredited facilities
- pH 4, 7, and 10 are color coded for easy identification
- Certified traceable to NIST
Specifications
7732-18-5,877-24-7 | |
Yes | |
pH Meter and Electrode | |
Liquid | |
1 L | |
1 L Natural Poly | |
Certified |
Colorless | |
Calibration | |
4.00 ±0.01 | |
VeriSpec | |
pH Buffer | |
ISO 17025/17034 |
ISO 17025/ISO 17034