missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ricca VeriSpec™ pH Buffer, Certified Reference Material

Ricca's VeriSpec™ ISO 17025/17034 pH Buffer Certified Reference Materials provide the highest level of quality assurance. A certificate of analysis providing the specified property value, uncertainty, and traceability is available with every bottle.

Supplier:  Ricca VeriSpec™ RV1600001A

 View more versions of this product

Catalog No. RV1600001A


Explore available promotions

Includes: 1L pH Buffer (10 pH)

Description

Description

  • No formaldehyde or mercury compounds as preservatives
  • No chromium compounds as colorants
  • Manufactured and tested in ISO 17025/17034 accredited facilities
  • pH 4, 7, and 10 are color coded for easy identification
  • Certified traceable to NIST
Specifications

Specifications

7732-18-5,497-19-8,1310-73-2,144-55-8
Yes
pH Meter and Electrode
Liquid
1 L
1 L Natural Poly
Certified
Colorless
Calibration
10.00 ±0.01
VeriSpec
pH Buffer
ISO 17025/17034
Videos
SDS
Documents

Documents

Promotions

Promotions

ISO 17025/ISO 17034