Learn More
Phenanthrene-9-boronic acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5254603
Description
Phenanthrene-9-boronic acid as a reactant is involved in the synthesis of large polycyclic aromatic hydrocarbons (PAHs) and electron rich benzo[g,h,i]perylenes. It is also used for preparation of FLAP protein inhibitors as anti-inflammatory agents and aminoisoquinolinones as PARP-2 selective inhibitors. As a reactant it is involved in homocoupling reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhenanthrene-9-boronic acid as a reactant is involved in the synthesis of large polycyclic aromatic hydrocarbons (PAHs) and electron rich benzo[g,h,i]perylenes. It is also used for preparation of FLAP protein inhibitors as anti-inflammatory agents and aminoisoquinolinones as PARP-2 selective inhibitors. As a reactant it is involved in homocoupling reactions.
Solubility
Soluble in DMSO.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Phenanthrene-9-boronic acid | |
254°C to 256°C | |
MFCD00143524 | |
9-phenanthreneboronic acid, 9-phenanthracenylboronic acid, phenanthrene-9-boronic acid, 9-phenanthrenylboronic acid, 9-phenanthrene boronic acid, 9-phenanthreneboronicacid, 9-phenanthrylboronic acid, boronic acid, 9-phenanthrenyl, phenanthracene-9-boronic acid, phenanthreneboronicacid | |
JCDAUYWOHOLVMH-UHFFFAOYSA-N | |
phenanthren-9-ylboronic acid | |
11775704 | |
97% |
68572-87-2 | |
C14H11BO2 | |
1 g | |
Soluble in DMSO. | |
OB(O)C1=CC2=CC=CC=C2C2=CC=CC=C12 | |
222.05 | |
222.05 |
Chemical Identifiers
68572-87-2 | |
222.05 | |
JCDAUYWOHOLVMH-UHFFFAOYSA-N | |
11775704 | |
OB(O)C1=CC2=CC=CC=C2C2=CC=CC=C12 |
C14H11BO2 | |
MFCD00143524 | |
9-phenanthreneboronic acid, 9-phenanthracenylboronic acid, phenanthrene-9-boronic acid, 9-phenanthrenylboronic acid, 9-phenanthrene boronic acid, 9-phenanthreneboronicacid, 9-phenanthrylboronic acid, boronic acid, 9-phenanthrenyl, phenanthracene-9-boronic acid, phenanthreneboronicacid | |
phenanthren-9-ylboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only