Learn More
Phenazine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals AC130150050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Phenazine | |
92-82-0 | |
Yellow to Brown | |
Authentic | |
Glass bottle | |
MFCD00005023 | |
23,223 | |
dibenzopyrazine, azophenylene, dibenzoparadiazine, acridizine, 9,10-diazaanthracene, dibenzo-p-diazine, unii-2jhr6k463w, phenazine, dibenzo b,e pyrazine, acmc-209rhs | |
PCNDJXKNXGMECE-UHFFFAOYSA-N | |
phenazine | |
4757 | |
180.21 | |
Crystalline Powder |
98% | |
174.0°C to 177.0°C | |
>360.0°C | |
97.5% min. (HPLC) | |
C12H8N2 | |
5 g | |
15,7331 | |
Solubility in water: insoluble. Other solubilities: partially soluble in alcohol and ether, moderately soluble in benzene | |
C1=CC=C2C(=C1)N=C3C=CC=CC3=N2 | |
180.21 | |
CHEBI:36674 | |
98% |
Chemical Identifiers
92-82-0 | |
180.21 | |
PCNDJXKNXGMECE-UHFFFAOYSA-N | |
4757 | |
phenazine |
C12H8N2 | |
MFCD00005023 | |
dibenzopyrazine, azophenylene, dibenzoparadiazine, acridizine, 9,10-diazaanthracene, dibenzo-p-diazine, unii-2jhr6k463w, phenazine, dibenzo b,e pyrazine, acmc-209rhs | |
CHEBI:36674 | |
C1=CC=C2C(=C1)N=C3C=CC=CC3=N2 |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Suspected of causing genetic defects.
GHS P Statement:
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 202-193-9
RTECSNumber : SG1360000
TSCA : TSCA
RUO – Research Use Only