missing translation for 'onlineSavingsMsg'
Learn More

Phenethylamine, 99%

CAS: 64-04-0 | C8H11N | 121.18 g/mol

$60.31 - $104.32

Chemical Identifiers

CAS 64-04-0
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00008184
InChI Key BHHGXPLMPWCGHP-UHFFFAOYSA-N
Synonym phenethylamine, 2-phenylethylamine, benzeneethanamine, phenylethylamine, 2-phenethylamine, beta-phenylethylamine, beta-phenethylamine, 1-amino-2-phenylethane, 2-aminoethyl benzene, beta-aminoethylbenzene
PubChem CID 1001
ChEBI CHEBI:18397
IUPAC Name 2-phenylethanamine
SMILES C1=CC=C(C=C1)CCN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC156491000
SDS View Documents
Thermo Scientific Chemicals
156491000
100 g Glass bottle
Each for $60.31
Only null left
 
AC156492500
SDS View Documents
Thermo Scientific Chemicals
156492500
250 g Glass bottle
Each for $104.32
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

64-04-0
121.18
BHHGXPLMPWCGHP-UHFFFAOYSA-N
1001
2-phenylethanamine
C8H11N
MFCD00008184
phenethylamine, 2-phenylethylamine, benzeneethanamine, phenylethylamine, 2-phenethylamine, beta-phenylethylamine, beta-phenethylamine, 1-amino-2-phenylethane, 2-aminoethyl benzene, beta-aminoethylbenzene
CHEBI:18397
C1=CC=C(C=C1)CCN

Specifications

64-04-0
Colorless to Yellow
200.0°C to 202.0°C
Authentic
Glass bottle
1.5320 to 1.5340
MFCD00008184
12, 1096
0.985
phenethylamine, 2-phenylethylamine, benzeneethanamine, phenylethylamine, 2-phenethylamine, beta-phenylethylamine, beta-phenethylamine, 1-amino-2-phenylethane, 2-aminoethyl benzene, beta-aminoethylbenzene
BHHGXPLMPWCGHP-UHFFFAOYSA-N
2-phenylethanamine
1001
CHEBI:18397
99%
Phenethylamine
-60.0°C
0.9850g/mL
81°C
98.5% min. (GC)
C8H11N
C6H5CH2CH2NH2
100 g
09,363
15, 7339
Solubility in water: 63g/L (25°C). Other solubilities: soluble in ethanol, diethyl ether
C1=CC=C(C=C1)CCN
121.18
2.4 mm2/s (23°C)
121.18
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 200-574-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only