missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenetole, 98+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 221491000

Catalog No. AC221491000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Phenetole
103-73-1
169.0°C to 170.0°C
Authentic
Glass bottle
1.5055 to 1.5075
MFCD00009090
06, 140
15, 7343
Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and carbon tetrachloride
CCOC1=CC=CC=C1
122.17
CHEBI:67129
98+%
99%
0.9600g/mL
57°C
98% min. (GC)
C8H10O
C6H5OC2H5
100mL
0.96
phenetole, benzene, ethoxy, ethyl phenyl ether, phenetol, phenyl ethyl ether, phenoxyethane, a phenoxyethane, ether, ethyl phenyl, unii-rb8lu2c57f
DLRJIFUOBPOJNS-UHFFFAOYSA-N
ethoxybenzene
7674
122.17
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.

GHS Signal Word: Warning

EINECSNumber : 203-139-7

RTECSNumber : SI7700000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only